accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 629-59-4

IUPAC Name: tetradecane
Synonyms: tetradecane tetradekan ccris 715 N-tetradecane
Molecular Weight (g/mol): 198.39
Molecular Formula: C14H30
InChi Key: BGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (13)
  • (12)
Boiling Point 
  • (1)
  • (6)
  • (5)
  • (5)
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (6)
Physical Form 
  • (19)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)
  • (3)
  • (9)
  • (13)
  • (12)
  • (1)
  • (6)
  • (5)
  • (5)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (6)
  • (19)
  • (2)
  • (2)
  • (5)
  • (1)