accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62922-45-6

ClH3NClCl

CAS RN 62922-45-6

IUPAC Name: 2,5-dichloropentan-1-aminium chloride
Synonyms: 2,5-dichloropentan-1-aminium chloride
Molecular Weight (g/mol): 192.51
Molecular Formula: C5H12Cl3N
InChi Key: ATGSLQBVSZNJMT-UHFFFAOYNA-N
SMILES: [Cl-].[NH3+]CC(Cl)CCCCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)