Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloroamylamine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. D279725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D279725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D279725G Supplier TCI America Supplier No. D279725G

missing translation for 'validMainframeMsg'

Specifications

CAS 62922-45-6
Melting Point 160°C
Color White-Yellow
Molecular Formula C5H12Cl3N
Quantity 25 g
MDL Number MFCD00012877
Synonym 2,5-Dichloropentylamine Hydrochloride
InChI Key ATGSLQBVSZNJMT-UHFFFAOYNA-N
SMILES [Cl-].[NH3+]CC(Cl)CCCCl
IUPAC Name 2,5-dichloropentan-1-aminium chloride
Molecular Weight (g/mol) 192.51
PubChem CID 2737572
Formula Weight 192.51
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroamylamine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.