accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6293-87-4

IUPAC Name: 2-(2-nitrophenyl)hydrazinium chloride
Synonyms: 2-(2-nitrophenyl)hydrazinium chloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C6H8ClN3O2
InChi Key: XCUBVSAYUSFHNN-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]NC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results