Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. N023125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N023125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N023125G Supplier TCI America Supplier No. N023125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6293-87-4
Molecular Formula C6H8ClN3O2
Molecular Weight (g/mol) 189.60
MDL Number MFCD00060205
InChI Key XCUBVSAYUSFHNN-UHFFFAOYSA-N
PubChem CID 5743520
IUPAC Name 2-(2-nitrophenyl)hydrazinium chloride
SMILES [Cl-].[NH3+]NC1=CC=CC=C1[N+]([O-])=O

Specifications

Quantity 25 g
UN Number 1325
Formula Weight 189.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenylhydrazine Hydrochloride
RTECSNumber MV8230000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.