accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOHO

CAS RN 62935-72-2

IUPAC Name: 2-(7-methoxy-2-oxo-2H-chromen-4-yl)acetic acid
Synonyms: (7-methoxy-2-oxochromen-4-yl)acetic acid
Molecular Weight (g/mol): 234.21
Molecular Formula: C12H10O5
InChi Key: ZEKAXIFHLIITGV-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
Technique 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)