Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-methoxycoumarin-4-acetic acid UV-VIS Internal Standard in Water, 41 ppm, SPEX CertiPrep™
SDP

Catalog No. UV323W Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UV323W 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UV323W Supplier SPEX CertiPrep Supplier No. UV323W

missing translation for 'validMainframeMsg'

UV-VIS internal standards allow the user to accurately measure the concentration of their samples by directly comparing the absorbance of their compounds against that of the internal standard.

SPEX Certiprep™ offers eight different standards that cover a wide range of wavelengths (270 to 627 nm). After the user takes a UV-VIS spectrum of their sample, they can find a region in the spectrum that doesn’t absorb, and then select the corresponding internal standard, and rerun the spectrum.

Specifications

Chemical Name or Material 7-methoxycoumarin-4-acetic acid
Packaging Ampoule
CAS 62935-72-2
Molecular Formula C12H10O5
Concentration 41 ppm
Technique UV/Vis, Spectrometry, LCMS and HPLC
Color Colorless
Synonym UV-323 nm internal standard
Solubility Soluble in water
Type UV Internal Standard
Molecular Weight (g/mol) 234.2
Quantity 3.5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.