accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62936-23-6

IUPAC Name: 3-chloro-4-hydroxy-5-methoxybenzoic acid
Synonyms: 3-chloro-4-hydroxy-5-methoxybenzoic acid
Molecular Weight (g/mol): 202.59
Molecular Formula: C8H7ClO4
InChi Key: XBRYEHVBBMSSCG-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(Cl)=C1O)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results