accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6295-35-8

CH3OOOCH3

CAS RN 6295-35-8

IUPAC Name: 6-methoxy-4-methyl-2H-chromen-2-one
Synonyms: 6-methoxy-4-methylchromen-2-one
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: KNGPIBWCWOQBEK-UHFFFAOYSA-N
SMILES: COC1=CC=C2OC(=O)C=C(C)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results