Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. M13985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13985G 5 g
M139825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13985G Supplier TCI America Supplier No. M13985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6295-35-8
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00635101
InChI Key KNGPIBWCWOQBEK-UHFFFAOYSA-N
PubChem CID 223821
IUPAC Name 6-methoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=C(C=C2)OC

Specifications

Melting Point 164°C
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-4-methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.