accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6296-42-0

IUPAC Name: (2-chloroethyl)urea
Synonyms: 2-chloroethylurea
Molecular Weight (g/mol): 122.55
Molecular Formula: C3H7ClN2O
InChi Key: BITBMHVXCILUEX-UHFFFAOYSA-N
SMILES: NC(=O)NCCCl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results