Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethylurea 98.0+%, TCI America™
SDP

Catalog No. C017325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C017325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C017325G Supplier TCI America Supplier No. C017325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6296-42-0
Molecular Formula C3H7ClN2O
Molecular Weight (g/mol) 122.552
MDL Number MFCD00059861
InChI Key BITBMHVXCILUEX-UHFFFAOYSA-N
PubChem CID 240817
IUPAC Name 2-chloroethylurea
SMILES C(CCl)NC(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 122.55
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethylurea
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.