accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO(S)OH(R)OHOOCH3CH3

CAS RN 62961-64-2

IUPAC Name: 1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Synonyms: 1,4-diisopropyl (2R,3S)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 234.25
Molecular Formula: C10H18O6
InChi Key: XEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILES: CC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)