Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diisopropyl D-tartrate, 98%

Catalog No. AAA1736214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1736214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1736214 Supplier Thermo Scientific Chemicals Supplier No. A1736214

missing translation for 'validMainframeMsg'

CAS: 62961-64-2 | C10H18O6 | 234.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62961-64-2
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00008876
InChI Key XEBCWEDRGPSHQH-YUMQZZPRSA-N
Synonym diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
PubChem CID 112972
IUPAC Name dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

Specifications

Density 1.117
Boiling Point 150°C to 151°C (11 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.44
Quantity 25 g
Beilstein 5265431
Optical Rotation −16° (Neat)
Formula Weight 234.25
Percent Purity 98%
Chemical Name or Material (-)-Diisopropyl D-tartrate
EINECSNumber 263-771-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.