accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6297-03-6

IUPAC Name: 1-(octadecyloxy)octadecane
Synonyms: distearyl ether
Molecular Weight (g/mol): 522.99
Molecular Formula: C36H74O
InChi Key: HBXWUCXDUUJDRB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results