accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOCH2OOH2COOCH2

CAS RN 6299-73-6

IUPAC Name: 1,2,3-tris(prop-2-en-1-yl) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: 1,2,3-tris(prop-2-en-1-yl) 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol): 312.32
Molecular Formula: C15H20O7
InChi Key: PLCFYBDYBCOLSP-UHFFFAOYSA-N
SMILES: OC(CC(=O)OCC=C)(CC(=O)OCC=C)C(=O)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results