Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl Citrate 90.0+%, TCI America™
SDP

Catalog No. C10385ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10385ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10385ML Supplier TCI America Supplier No. C10385ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6299-73-6
Molecular Formula C15H20O7
Molecular Weight (g/mol) 312.318
MDL Number MFCD00059327
InChI Key PLCFYBDYBCOLSP-UHFFFAOYSA-N
Synonym Citric Acid Triallyl Ester
PubChem CID 80534
IUPAC Name tris(prop-2-enyl) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES C=CCOC(=O)CC(CC(=O)OCC=C)(C(=O)OCC=C)O

Specifications

Boiling Point 208°C
Quantity 5 mL
Formula Weight 312.32
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Triallyl Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.