CAS RN 6299-87-2
IUPAC Name:
6-oxo-3,6-dihydropyrimidine-4-carboxylic acid
Synonyms:
6-oxo-3H-pyrimidine-4-carboxylic acid
Molecular Weight (g/mol):
140.1
Molecular Formula:
C5H4N2O3
InChi Key:
QYGHXDAYBIFGKI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=O)N=CN1
1
–
1
of
1
results
1
6-Hydroxy-4-pyrimidinecarboxylic Acid 98.0+%, TCI America™
CAS: 6299-87-2 Molecular Formula: C5H4N2O3 Molecular Weight (g/mol): 140.10 MDL Number: MFCD00023267 InChI Key: QYGHXDAYBIFGKI-UHFFFAOYSA-N Synonym: 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid PubChem CID: 239692 IUPAC Name: 6-oxo-3,6-dihydropyrimidine-4-carboxylic acid SMILES: OC(=O)C1=CC(=O)N=CN1