accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOO

CAS RN 63-25-2

IUPAC Name: naphthalen-1-yl N-methylcarbamate
Synonyms: carbaryl carbaryl product caswell no. 160 carbaril coophavet brand panam denapon fido's freeitch fido free-itch sépou karbaspray
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: CVXBEEMKQHEXEN-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results