accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 630-04-6

IUPAC Name: hentriacontane
Synonyms: hentriacontane
Molecular Weight (g/mol): 436.85
Molecular Formula: C31H64
InChi Key: IUJAMGNYPWYUPM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)