Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hentriacontane 95.0+%, TCI America™
SDP

Catalog No. H00121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00121G 1 g
H001210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00121G Supplier TCI America Supplier No. H00121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 630-04-6
Molecular Formula C31H64
Molecular Weight (g/mol) 436.85
MDL Number MFCD00048745
InChI Key IUJAMGNYPWYUPM-UHFFFAOYSA-N
Synonym n-hentriacontane, untriacontane, unii-6sdg640hl3, hentriacontan, candelilla wax, candellila wax, waxes, candelilla, triacontane, methyl, euphorbia cerifera wax, acmc-1b2c1
PubChem CID 12410
ChEBI CHEBI:5659
IUPAC Name hentriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 66°C
Color White
Boiling Point 180°C
Quantity 1 g
Formula Weight 436.85
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hentriacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.