accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 630-07-9

H3CCH3

CAS RN 630-07-9

IUPAC Name: pentatriacontane
Synonyms: N-pentatriacontane
Molecular Weight (g/mol): 492.96
Molecular Formula: C35H72
InChi Key: VHQQPFLOGSTQPC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results