accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SeHN

CAS RN 630-10-4

IUPAC Name: [(C-selanylcarbonimidoyl)amino]-λ¹-selanyl
Synonyms: selenourea
Molecular Weight (g/mol): 122.02
Molecular Formula: CH3N2Se
InChi Key: ZUCFIOXGLBMWHV-UHFFFAOYSA-N
SMILES: NC([Se])=N
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (4)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)