Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenourea, 99%

Catalog No. AAB2144006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2144006 5 g
AAB2144003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2144006 Supplier Thermo Scientific Chemicals Supplier No. B2144006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 630-10-4 | CH3N2Se | 122.02 g/mol

Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

Solubility
Soluble in water, alcohol and ether.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-10-4
Molecular Formula CH3N2Se
Molecular Weight (g/mol) 122.02
MDL Number MFCD00008065
InChI Key ZUCFIOXGLBMWHV-UHFFFAOYSA-N
Synonym selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
PubChem CID 6327594
IUPAC Name $l^{1}-selanylmethanimidamide
SMILES C(=N)(N)[Se]

Specifications

Melting Point ∼210°C (decomposition)
Assay Percent Range 99+% metals basis
Quantity 5 g
UN Number UN3283
Sensitivity Air, light, and moisture sensitive
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 123.02
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Selenourea
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber 211-129-9
RTECSNumber YU1820000
TSCA Yes
storageNote1 Air and Moisture Sensitive
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.