accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 630-20-6

IUPAC Name: 1,1,1,2-tetrachloroethane
Synonyms: tetrachloroethane 1,1,1,2-tetrachloraethan C2H2cl4 ccris 577 rcra waste number U208
Molecular Weight (g/mol): 167.84
Molecular Formula: C2H2Cl4
InChi Key: QVLAWKAXOMEXPM-UHFFFAOYSA-N
SMILES: ClCC(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results