Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2-Tetrachloroethane 99.0+%, TCI America™
SDP

Catalog No. T069510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T069510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T069510G Supplier TCI America Supplier No. T069510G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 630-20-6
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
MDL Number MFCD00000807
InChI Key QVLAWKAXOMEXPM-UHFFFAOYSA-N
Synonym ethane, 1,1,1,2-tetrachloro, rcra waste number u208, 1,1,1,2-tetrachlorethane, rcra waste no. u208, unii-0s5mke574x, ccris 577, 1,1,1,2-tetrachloroethane, analytical standard, tetrachloroethane, 1,1,1,2, ethyl, 1,2,2,2-tetrachloro, alpha,beta,beta,beta-tetrachloroethyl
PubChem CID 12418
ChEBI CHEBI:34024
IUPAC Name 1,1,1,2-tetrachloroethane
SMILES C(C(Cl)(Cl)Cl)Cl

Specifications

Color Colorless
Boiling Point 131°C
Quantity 10 g
UN Number 2810
Formula Weight 167.84
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2-Tetrachloroethane
EINECSNumber (2)-0056
RTECSNumber KI8450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.