CAS RN 630-56-8
IUPAC Name:
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl hexanoate
Synonyms:
proge hydroxyprogesterone caproate proluton depot lutate primolut depot hydroxyprogesterone hexanoate hylutin luteocrin hormofort oxyprogesterone caproate
Molecular Weight (g/mol):
428.61
Molecular Formula:
C27H40O4
InChi Key:
DOMWKUIIPQCAJU-LJHIYBGHSA-N
SMILES:
CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
1
–
1
of
1
results
1
Promotions Available
Hydroxyprogesterone Caproate, USP, 97-103%, Spectrum™ Chemical
Hydroxyprogesterone Caproate, USP, 97-103%, Spectrum™ Chemical