accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHN(S)CHOO

CAS RN 63039-48-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}pent-4-ynoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]pent-4-ynoate
Molecular Weight (g/mol): 212.23
Molecular Formula: C10H14NO4
InChi Key: AMKHAJIFPHJYMH-ZETCQYMHSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC#C)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results