Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-propargylglycine 97.0+%, TCI America™
SDP

Catalog No. B40071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40071G Supplier TCI America Supplier No. B40071G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63039-48-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD01320855
InChI Key AMKHAJIFPHJYMH-ZETCQYMHSA-N
Synonym boc-l-propargylglycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-pra-oh, boc-propargyl-gly-oh, n-boc-2-propargyl-l-glycine, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-propargylglycine, boc-l-propargyl glycine
PubChem CID 2734488
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O

Specifications

Melting Point 90°C
Color Yellow
Quantity 1 g
Formula Weight 213.23
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-propargylglycine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.