accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6304-24-1

IUPAC Name: 2-(2-methylpropyl)pyridine
Synonyms: 2-(2-methylpropyl)pyridine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: BBVSPSDWPYWMOR-UHFFFAOYSA-N
SMILES: CC(C)CC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results