accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOClI

CAS RN 63059-29-0

IUPAC Name: 2-iodobenzene-1-sulfonyl chloride
Synonyms: 2-iodobenzenesulfonyl chloride
Molecular Weight (g/mol): 302.51
Molecular Formula: C6H4ClIO2S
InChi Key: ZPNWNLNPDRYULE-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)