Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. I08115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08115G 5 g
I08111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08115G Supplier TCI America Supplier No. I08115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63059-29-0
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00833336
InChI Key ZPNWNLNPDRYULE-UHFFFAOYSA-N
PubChem CID 627400
IUPAC Name 2-iodobenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC=C1I

Specifications

Melting Point 52°C
Boiling Point 135°C
Quantity 5 g
UN Number 3261
Formula Weight 302.51
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.