accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONHOO

CAS RN 63059-33-6

IUPAC Name: 2-[(butylcarbamoyl)amino]acetate
Synonyms: 2-[(butylcarbamoyl)amino]acetate
Molecular Weight (g/mol): 173.19
Molecular Formula: C7H13N2O3
InChi Key: KGWHMDCQVDMTNZ-UHFFFAOYSA-M
SMILES: CCCCNC(=O)NCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results