Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Butylhydantoic Acid 98%, Thermo Scientific™

Catalog No. AC403550250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC403550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC403550250 Supplier Thermo Scientific Chemicals Supplier No. 403550250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63059-33-6
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
InChI Key KGWHMDCQVDMTNZ-UHFFFAOYSA-N
Synonym 5-butylhydantoic acid, n-butylamino carbonyl glycine, glycine, n-butylamino carbonyl, 2-3-butylureido acetic acid, n-butyl-n'-carboxymethyl urea, 2-butylamino carbonylamino acetic acid, butylcarbamoyl amino acetic acid, n-butylcarbamoyl glycine
PubChem CID 112991
IUPAC Name 2-(butylcarbamoylamino)acetic acid
SMILES CCCCNC(=O)NCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3(CH2)3NHCONHCH2CO2H
Quantity 25 g
Formula Weight 174.2
Percent Purity ≥97.5%
Physical Form Fine Powder
Chemical Name or Material 5-Butylhydantoic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.