accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 6306-07-6

IUPAC Name: 1,2-dihydroacenaphthylen-1-ol
Synonyms: 1,2-dihydroacenaphthylen-1-ol
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: MXUCIEHYJYRTLT-UHFFFAOYNA-N
SMILES: OC1CC2=C3C1=CC=CC3=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results