accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FN

CAS RN 63069-50-1

IUPAC Name: 4-amino-3-fluorobenzonitrile
Synonyms: 4-amino-3-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: RLMBRRQWBTWGMB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results