CAS RN 63074-08-8
IUPAC Name:
hydrogen 6,7-dimethoxy-2-[4-(oxolane-2-carbonyl)piperazin-1-yl]quinazolin-4-amine chloride
Synonyms:
terazosin
Molecular Weight (g/mol):
423.9
Molecular Formula:
C19H26ClN5O4
InChi Key:
IWSWDOUXSCRCKW-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1CCCO1
1
–
1
of
1
results
1
R&D Systems™ Terazosin hydrochloride
CAS: 63074-08-8 Molecular Formula: C19H26ClN5O4