accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63076-51-7

IUPAC Name: cyclopentylboronic acid
Synonyms: cyclopentylboronic acid
Molecular Weight (g/mol): 113.95
Molecular Formula: C5H11BO2
InChi Key: VTTDFSNKIMAQTB-UHFFFAOYSA-N
SMILES: OB(O)C1CCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results