Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C24425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2442-5G 5 g
C24421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2442-5G Supplier TCI America Supplier No. C24425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
MDL Number MFCD01074541
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid

Specifications

Melting Point 126°C
Quantity 5 g
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792
SMILES OB(O)C1CCCC1
Molecular Weight (g/mol) 113.95
Formula Weight 113.95
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.