CAS RN 631-31-2
CAS RN 631-31-2
IUPAC Name:
2-ethyl-2-methylbutanedioic acid
Synonyms:
2-ethyl-2-methyl-succinic acid
Molecular Weight (g/mol):
160.17
Molecular Formula:
C7H12O4
InChi Key:
FDYJJKHDNNVUDR-UHFFFAOYNA-N
SMILES:
CCC(C)(CC(O)=O)C(O)=O
1
–
1
of
1
results
1
2-Ethyl-2-Methylsuccinic Acid, MP Biomedicals™
CAS: 631-31-2 Molecular Formula: C7H12O4 Molecular Weight (g/mol): 160.17 MDL Number: MFCD00004201 InChI Key: FDYJJKHDNNVUDR-UHFFFAOYNA-N Synonym: Isopimelic Acid,α-Ethyl methyl succinic acid