Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-Methylsuccinic Acid, MP Biomedicals™

Catalog No. MP52058676 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP52058676 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP52058676 Supplier MP Biomedicals, Inc Supplier No. 05205867.6
Add to Cart
Add to Cart

2-Ethyl-2-Methylsuccinic Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-31-2
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004201
InChI Key FDYJJKHDNNVUDR-UHFFFAOYNA-N
Synonym Isopimelic Acid, α-Ethyl methyl succinic acid

Specifications

Melting Point 100°C to 102°C
Color White
Quantity 50 mg
Percent Purity >98%
Physical Form Solid
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.