accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiOSiOHSiMeMeMeMeMeMeMen

CAS RN 63148-57-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (CH4OSi)n
InChi Key: QNWOFLWXQGHSRH-UHFFFAOYSA-N
SMILES: C[SiH](O[Si](C)(C)C)O[Si](C)(C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)