Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methylhydrosiloxane)

Catalog No. 17620650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-620-650G 50 g
17-620-6250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-620-650G Supplier Sigma Aldrich Supplier No. 17620650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is an environmentally benign reducing agent and has been enhanced for catalytic efficiency. Click here for more information.Harnessing the reactivity of poly(methylhydrosiloxane) for the reduction and cyclization of biomass to high-value products. Used in greener amine synthesis by reductive amination as an alternative to borohydrides.Synthesis of substituted amines and isoindolinones: catalytic reductive amination using abundantly available AlCl3/PMHSAmine Synthesis through Mild Catalytic Hydrosilylation of Imines using Polymethylhydroxysiloxane and [RuCl2(arene)]2 Catalysts

Specifications

CAS 63148-57-2
Density 1.006 g/mL (at 25°C)
Refractive Index n20/D 1.398
Linear Formula (CH3)3 SiO[(CH3)HSiO]nSi(CH3)3
MDL Number MFCD00084478
Quantity 50 g
Synonym Methylhydrogensiloxane polymer; PMHS
Molecular Weight (g/mol) 1,700-3,200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.