accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 63148-58-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)