accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHON

CAS RN 63149-33-7

IUPAC Name: 8-hydroxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Synonyms: 8-hydroxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Molecular Weight (g/mol): 217.27
Molecular Formula: C13H15NO2
InChi Key: NRZXBDYODHLZBF-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=C2CCCN3CCCC1=C23
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)