Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyjulolidine-9-carboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. H12255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12255G Supplier TCI America Supplier No. H12255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63149-33-7
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.268
MDL Number MFCD00192477
InChI Key NRZXBDYODHLZBF-UHFFFAOYSA-N
Synonym 8-hydroxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline-9-carbaldehyde, 8-hydroxyjulolidine-9-aldehyde, 9-formyl-8-hydroxyjulolidine, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo ij quinolizine-9-carboxaldehyde, 8-hydroxyjulolidine-9-carboxaldehyde, 9-formyl-8-julolidinol, 1h,5h-benzo ij quinolizine-9-carboxaldehyde, 2,3,6,7-tetrahydro-8-hydroxy, 2,3,6,7-tetrahydro-8-hydroxy-1h,5h-benzo i,j quinolizine-9-carboxaldehyde, 8-hydroxy-2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline-9-carbaldehyde
PubChem CID 113099
SMILES C1CC2=C3C(=C(C(=C2)C=O)O)CCCN3C1

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxyjulolidine-9-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.