accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHHO(E)

CAS RN 63177-57-1

IUPAC Name: methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Synonyms: methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: BQCNSTFWSKOWMA-GORDUTHDSA-N
SMILES: COC(=O)\C=C\C1=CC(O)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results