CAS RN 6319-96-6
CAS RN 6319-96-6
IUPAC Name:
2-(2-chloroacetamido)acetic acid
Synonyms:
(2-chloroacetamido)acetic acid
Molecular Weight (g/mol):
151.55
Molecular Formula:
C4H6ClNO3
InChi Key:
DLDTUYIGYMNERN-UHFFFAOYSA-N
SMILES:
OC(=O)CNC(=O)CCl
1
–
1
of
1
results
1
N-Chloroacetylglycine 99.0+%, TCI America™

CAS: 6319-96-6 Molecular Formula: C4H6ClNO3 Molecular Weight (g/mol): 151.546 MDL Number: MFCD00021733 InChI Key: DLDTUYIGYMNERN-UHFFFAOYSA-N Synonym: n-chloroacetylglycine,n-chloroacetyl glycine,2-2-chloroacetyl amino acetic acid,2-chloroacetamido acetic acid,2-2-chloroacetamido acetic acid,chloracetyl-glycin,chloroac-gly-oh,chloroacetyl glycine,acmc-209nds,chloroacetylglycine PubChem CID: 233485 IUPAC Name: 2-[(2-chloroacetyl)amino]acetic acid SMILES: C(C(=O)O)NC(=O)CCl