Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetylglycine 99.0+%, TCI America™
SDP

Catalog No. C098025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C098025G 25 g
C09801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C098025G Supplier TCI America Supplier No. C098025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6319-96-6
Molecular Formula C4H6ClNO3
Molecular Weight (g/mol) 151.546
MDL Number MFCD00021733
InChI Key DLDTUYIGYMNERN-UHFFFAOYSA-N
Synonym n-chloroacetylglycine, n-chloroacetyl glycine, 2-2-chloroacetyl amino acetic acid, 2-chloroacetamido acetic acid, 2-2-chloroacetamido acetic acid, chloracetyl-glycin, chloroac-gly-oh, chloroacetyl glycine, acmc-209nds, chloroacetylglycine
PubChem CID 233485
IUPAC Name 2-[(2-chloroacetyl)amino]acetic acid
SMILES C(C(=O)O)NC(=O)CCl

Specifications

Melting Point 102°C
Color White
Quantity 25 g
Formula Weight 151.55
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.