accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 632-83-7

IUPAC Name: 1-bromo-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-bromoanthraquinone
Molecular Weight (g/mol): 287.11
Molecular Formula: C14H7BrO2
InChi Key: CXTPIHZYOGDSLV-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results